Acetic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals
Acetic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
1 L
4 L
Catalog number 035567.M4
also known as 035567-M4
Price (USD)/ Each
177.00
-
Add to cart
Quantity:
20 L
Request bulk or custom format
Price (USD)/ Each
177.00
Add to cart
Acetic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals
Catalog number035567.M4
Price (USD)/ Each
177.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAcetic acid
CAS64-19-7
Quantity20 L
Recommended StorageAmbient temperatures
EINECS Number200-580-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only