Ammonium hydroxide, 10% v/v aq. soln., Thermo Scientific Chemicals
Ammonium hydroxide, 10% v/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydroxide, 10% v/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 035575.AP
also known as 035575-AP
Price (USD)/ Each
37.65
Online exclusive
41.60 
Save 3.95 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.60 
Save 3.95 (10%)
Add to cart
Ammonium hydroxide, 10% v/v aq. soln., Thermo Scientific Chemicals
Catalog number035575.AP
Price (USD)/ Each
37.65
Online exclusive
41.60 
Save 3.95 (10%)
-
Add to cart
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration v/v10% v/v aq. soln.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only