Ammonium molybdate (di), 10% w/v aq. soln., Thermo Scientific Chemicals
Ammonium molybdate (di), 10% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium molybdate (di), 10% w/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 035579.AP
Price (USD)/ Each
78.65
Online exclusive
87.20 
Save 8.55 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
87.20 
Save 8.55 (10%)
Add to cart
Ammonium molybdate (di), 10% w/v aq. soln., Thermo Scientific Chemicals
Catalog number035579.AP
Price (USD)/ Each
78.65
Online exclusive
87.20 
Save 8.55 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAmmonium molybdate (di)
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only