Barium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Barium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number 035582.K7
also known as 035582-K7
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
Add to cart
Barium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Catalog number035582.K7
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10361-37-2
IUPAC Namebarium(2+) dichloride
Molecular FormulaBaCl2
InChI KeyWDIHJSXYQDMJHN-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay from Supplier's CofA0.0998 N - 0.1002 N
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only