Potassium hexacyanoferrate(II), 0.1N Standardized Solution, Thermo Scientific Chemicals
Potassium hexacyanoferrate(II), 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hexacyanoferrate(II), 0.1N Standardized Solution, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 L
Catalog number 035585.K2
also known as 035585-K2
Price (USD)/ Each
70.65
Online exclusive
77.80 
Save 7.15 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
77.80 
Save 7.15 (9%)
Add to cart
Potassium hexacyanoferrate(II), 0.1N Standardized Solution, Thermo Scientific Chemicals
Catalog number035585.K2
Price (USD)/ Each
70.65
Online exclusive
77.80 
Save 7.15 (9%)
-
Add to cart
Chemical Identifiers
CAS13943-58-3
IUPAC Nametetrapotassium ironhexacarbonitrile
Molecular FormulaC6H6FeK4N6
InChI KeyFIBBHPJORRWWPX-UHFFFAOYSA-N
SMILES[K+].[K+].[K+].[K+].[Fe].C#N.C#N.C#N.C#N.C#N.C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.1N Potassium hexacyanoferrate(II)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only