Potassium dichromate, 0.25N Standardized Solution, Thermo Scientific Chemicals
Potassium dichromate, 0.25N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium dichromate, 0.25N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035587.K2
also known as 035587-K2
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.50 
Save 7.85 (14%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.50 
Save 7.85 (14%)
Add to cart
Potassium dichromate, 0.25N Standardized Solution, Thermo Scientific Chemicals
Catalog number035587.K2
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.50 
Save 7.85 (14%)
-
Add to cart
Chemical Identifiers
CAS7778-50-9
IUPAC Namedipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2K2O7
InChI KeyKMUONIBRACKNSN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.25N (0.04167M) ±0.5% @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only