Potassium bromate/Potassium bromide, 0.1N Standardized Solution, Thermo Scientific Chemicals
Potassium bromate/Potassium bromide, 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium bromate/Potassium bromide, 0.1N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 035593.K2
Price (USD)/ Each
86.65
Online exclusive
95.60 
Save 8.95 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
95.60 
Save 8.95 (9%)
Add to cart
Potassium bromate/Potassium bromide, 0.1N Standardized Solution, Thermo Scientific Chemicals
Catalog number035593.K2
Price (USD)/ Each
86.65
Online exclusive
95.60 
Save 8.95 (9%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPotassium bromate/Potassium bromide
Health Hazard 1H350
Quantity1 L
Health Hazard 3P201-P202-P281-P308+P313-P501c
ColorColorless
View more
Potassium bromate/Potassium bromide is used as an analytical reagent for volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only