Thermo Scientific Chemicals

Silver nitrate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035601.AP
500 mL, Each
Thermo Scientific Chemicals

Silver nitrate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035601.AP
500 mL, Each
Quantity
Catalog number: 035601.AP
also known as 035601-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7761-88-8
IUPAC Name
silver(1+) nitrate
Molecular Formula
AgNO3
InChI Key
SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES
[Ag+].[O-][N+]([O-])=O
Appearance (Color)
Colorless
Assay from Supplier's CofA
0.1N
Form
Liquid

Description

Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text