Sulfuric acid, 6.0N Standardized Solution, Thermo Scientific Chemicals
Sulfuric acid, 6.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfuric acid, 6.0N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035610.K2
also known as 035610-K2
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
57.10 
Save 8.45 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
57.10 
Save 8.45 (15%)
Add to cart
Sulfuric acid, 6.0N Standardized Solution, Thermo Scientific Chemicals
Catalog number035610.K2
Price (USD)/ Each
48.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
57.10 
Save 8.45 (15%)
-
Add to cart
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
ConcentrationSulfuric acid, 6.0N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov