Hydrochloric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrochloric acid, 10% v/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035611.AP
also known as 035611-AP
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.30 
Save 3.65 (13%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.30 
Save 3.65 (13%)
Add to cart
Hydrochloric acid, 10% v/v aq. soln., Thermo Scientific Chemicals
Catalog number035611.AP
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.30 
Save 3.65 (13%)
-
Add to cart
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration v/vHydrochloric acid, 10% v/v aq. soln.
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only