Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 035614.K2
also known as 035614-K2
Price (USD)/ Each
70.65
Online exclusive
77.90 
Save 7.25 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
77.90 
Save 7.25 (9%)
Add to cart
Ammonium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Catalog number035614.K2
Price (USD)/ Each
70.65
Online exclusive
77.90 
Save 7.25 (9%)
-
Add to cart
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.998 N - 1.002 N
Appearance (Color)Colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only