Sodium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Sodium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 035616.AP
also known as 035616-AP
Price (USD)/ Each
31.65
Online exclusive
35.30 
Save 3.65 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
31.65
Online exclusive
35.30 
Save 3.65 (10%)
Add to cart
Sodium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Catalog number035616.AP
Price (USD)/ Each
31.65
Online exclusive
35.30 
Save 3.65 (10%)
-
Add to cart
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.1N Sodium chloride
Sodium chloride, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only