Oxalic acid, 10% w/v aq. soln., Thermo Scientific Chemicals
Oxalic acid, 10% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxalic acid, 10% w/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number 035617.K7
also known as 035617-K7
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
Add to cart
Oxalic acid, 10% w/v aq. soln., Thermo Scientific Chemicals
Catalog number035617.K7
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialOxalic acid
CAS144-62-7
Health Hazard 1H290-H318
Quantity4 L
Health Hazard 3P234-P280-P305+P351+P338-P310-P390
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only