Thermo Scientific Chemicals

Potassium dichromate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035618.AP
500 mL, Each
Thermo Scientific Chemicals

Potassium dichromate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035618.AP
500 mL, Each
Quantity
Catalog number: 035618.AP
also known as 035618-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7778-50-9
IUPAC Name
dipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular Formula
Cr2K2O7
InChI Key
KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES
[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
Concentration
0.1N

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text