Sodium hydroxide, 0.01N Standardized Solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.01N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, 0.01N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
20 L
Catalog number 035620.K7
also known as 035620-K7
Price (USD)/ Each
69.60
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
69.60
Add to cart
Sodium hydroxide, 0.01N Standardized Solution, Thermo Scientific Chemicals
Catalog number035620.K7
Price (USD)/ Each
69.60
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.01N Sodium hydroxide
MatrixWater
Sodium hydroxide, 0.01N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only