Thermo Scientific Chemicals

Oxalic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035622.K2
1 L, Each
Thermo Scientific Chemicals

Oxalic acid, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035622.K2
1 L, Each
Quantity
Catalog number: 035622.K2
Price (USD)
Quantity
-
Have Questions?
Assay from Supplier's CofA
0.0499 M - 0.0501 M
Form
Liquid

Description

Oxalic acid, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text