Sodium hydroxide, 0.05N Standardized Solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.05N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, 0.05N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
1 L
4 L
Catalog number 035623.M4
also known as 035623-M4
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
20 L
Request bulk or custom format
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
Add to cart
Sodium hydroxide, 0.05N Standardized Solution, Thermo Scientific Chemicals
Catalog number035623.M4
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
Name NoteStandardized Solution
CAS1310-73-2
Recommended StorageAmbient temperatures
EINECS Number215-185-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only