Sodium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Sodium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
1 L
4 L
Catalog number 035629.M4
also known as 035629-M4
Price (USD)/ Each
230.00
-
Add to cart
Quantity:
20 L
Request bulk or custom format
Price (USD)/ Each
230.00
Add to cart
Sodium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Catalog number035629.M4
Price (USD)/ Each
230.00
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA1.0N
Appearance (Color)Colorless
FormLiquid
Sodium hydroxide, 1.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only