Hydrochloric acid, 5.0N Standardized Solution, Thermo Scientific Chemicals
Hydrochloric acid, 5.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrochloric acid, 5.0N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
20 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035638.K2
also known as 035638-K2
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.20 
Save 7.55 (16%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.20 
Save 7.55 (16%)
Add to cart
Hydrochloric acid, 5.0N Standardized Solution, Thermo Scientific Chemicals
Catalog number035638.K2
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.20 
Save 7.55 (16%)
-
Add to cart
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
MatrixWater
Concentration5.0 N Hydrochloric acid
Hydrochloric acid, 5.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only