Thermo Scientific Chemicals

Sodium hydroxide, 40% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 035639.M4
20 L, Each
Thermo Scientific Chemicals

Sodium hydroxide, 40% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 035639.M4
20 L, Each
Quantity
Catalog number: 035639.M4
also known as 035639-M4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Assay from Supplier's CofA
40% w/v ? 2% w/v
Form
Liquid

Description

Sodium hydroxide, 40% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text