Thermo Scientific Chemicals

Sodium thiosulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035645.K7
4 L, Each
Thermo Scientific Chemicals

Sodium thiosulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035645.K7
4 L, Each
Quantity
Catalog number: 035645.K7
also known as 035645-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7772-98-7
IUPAC Name
disodium sulfanidesulfonate
Molecular Formula
Na2O3S2
InChI Key
AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S([S-])(=O)=O
Assay from Supplier's CofA
0.1N
Form
Liquid

Description

Sodium thiosulfate, 0.1N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text