Thermo Scientific Chemicals

Cerium(IV) sulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035650.K7
4 L, Each
Thermo Scientific Chemicals

Cerium(IV) sulfate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 035650.K7
4 L, Each
Quantity
Catalog number: 035650.K7
also known as 035650-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13590-82-4
IUPAC Name
λ⁴-cerium(4+) disulfate
Molecular Formula
CeO8S2
InChI Key
VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Assay from Supplier's CofA
0.1N
Appearance (Color)
Yellow
Form
Liquid

Description

Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text