Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035661.09
also known as 035661-09
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
Add to cart
Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number035661.09
Price (USD)/ Each
146.65
Online exclusive
163.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12035-72-2
IUPAC Namedisulfanylidenetrinickel
Molecular FormulaNi3S2
InChI KeyYGHCWPXPAHSSNA-UHFFFAOYSA-N
SMILESS=[Ni][Ni][Ni]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only