Silicon powder, crystalline, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Silicon powder, crystalline, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon powder, crystalline, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 035662.18
also known as 035662-18
Price (USD)/ Each
233.65
Online exclusive
259.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
233.65
Online exclusive
259.00 
Save 25.35 (10%)
Add to cart
Silicon powder, crystalline, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number035662.18
Price (USD)/ Each
233.65
Online exclusive
259.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only