Zinc phosphide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc phosphide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc phosphide, 99.9% (metals basis), Thermo Scientific Chemicals

Useful applications in photovoltaic cells. It is also employed as a laboratory chemical.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.5 g
10 g
Catalog number 035664.04
also known as 035664-04
Price (USD)/ Each
230.65
Online exclusive
256.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
230.65
Online exclusive
256.00 
Save 25.35 (10%)
Add to cart
Zinc phosphide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number035664.04
Price (USD)/ Each
230.65
Online exclusive
256.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-84-7
IUPAC Namezinc phosphide
Molecular FormulaH2P2Zn3
InChI KeyHOKBIQDJCNTWST-UHFFFAOYSA-N
SMILES[Zn].P=[Zn].P=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only