Thermo Scientific Chemicals

Barium iodide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035679.14
25 g, Each
Thermo Scientific Chemicals

Barium iodide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035679.14
25 g, Each
Quantity
Catalog number: 035679.14
also known as 035679-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13718-50-8
IUPAC Name
barium(2+) diiodide
Molecular Formula
BaI2
InChI Key
SGUXGJPBTNFBAD-UHFFFAOYSA-L
SMILES
[I-].[I-].[Ba++]
Particle Size
-10 Mesh
Appearance (Color)
White to yellow
Residual water
< 100 ppm
Total Metal Impurities
0.005% max.
Form
Beads

Description

Barium iodide is used to prepare organobarium compounds by reacting with alkyl potassium compounds. It is also involved in the preparation of active barium metal by reducing with lithium biphenyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text