Cadmium iodide, ultra dry, 99.9985% (metals basis), Thermo Scientific Chemicals
Cadmium iodide, ultra dry, 99.9985% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium iodide, ultra dry, 99.9985% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035683.06
also known as 035683-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7790-80-9
IUPAC Namecadmium(2+) diiodide
Molecular FormulaCdI2
InChI KeyOKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES[Cd++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBeads
Total Metal Impurities0.0015% max.
Particle Size-10 Mesh
Appearance (Color)White
Residual water< 100 ppm
Cadmium iodide is used in lithography, photography, electroplating and the manufacturing of phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only