Calcium bromide, ultra dry , 99.978% (metals basis), Thermo Scientific Chemicals
Calcium bromide, ultra dry , 99.978% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium bromide, ultra dry , 99.978% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 035684.03
also known as 035684-03
Price (USD)/ Each
43.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
43.60
Add to cart
Calcium bromide, ultra dry , 99.978% (metals basis), Thermo Scientific Chemicals
Catalog number035684.03
Price (USD)/ Each
43.60
-
Add to cart
Chemical Identifiers
CAS7789-41-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities0.022% max.
Residual water< 100 ppm
Particle Size-10 Mesh
FormBeads
CaBr2 is mainly used as a dense aqueous solution for drilling fluid. It is also used in neuroses medication, freezing mixtures, food preservatives, photography and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only