Thermo Scientific Chemicals

Cerium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035688.14
25 g, Each
Thermo Scientific Chemicals

Cerium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035688.14
25 g, Each
Quantity
Catalog number: 035688.14
also known as 035688-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-86-5
IUPAC Name
cerium(3+) trichloride
Molecular Formula
CeCl3
InChI Key
VYLVYHXQOHJDJL-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ce+3]
Total Rare Earth Oxide Impurities
0.1% max.
Appearance (Color)
White
Residual water
< 100 ppm
Form
Crystalline

Description

Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text