Thermo Scientific Chemicals

Cesium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035690.06
5 g, Each
Thermo Scientific Chemicals

Cesium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035690.06
5 g, Each
Quantity
Catalog number: 035690.06
also known as 035690-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-17-8
IUPAC Name
caesium(1+) chloride
Molecular Formula
ClCs
InChI Key
AIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES
[Cl-].[Cs+]
Appearance (Color)
White
Residual water
< 100 ppm
Form
Beads
Particle Size
-10 Mesh
Total Metal Impurities
0.002% max.

Description

It is used as a precursor for cesium metal, it act as a phase transfer catalyst, for molecular genetic applications used in preparing nucleic acids for subsequent hybridization or cloning experiments. It a useful compound for density gradient centrifugation, for fractionation of ribosomal subunits, proteins, glycoproteins, and viruses, evacuation of radio tubes, in the preparation of electrically conducting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text