Chromium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Chromium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 035691.22
also known as 035691-22
Price (USD)/ Each
272.65
Online exclusive
303.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
272.65
Online exclusive
303.00 
Save 30.35 (10%)
Add to cart
Chromium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number035691.22
Price (USD)/ Each
272.65
Online exclusive
303.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-73-7
IUPAC Namechromium(3+) hexahydrate trichloride
Molecular FormulaCl3CrH12O6
InChI KeyLJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. It is used as a catalyst, for organic synthesis, for manufacturing dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only