Thermo Scientific Chemicals

Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals

Catalog number: 035692.06
5 g, Each
Thermo Scientific Chemicals

Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals

Catalog number: 035692.06
5 g, Each
Quantity
Catalog number: 035692.06
also known as 035692-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-74-8
IUPAC Name
dysprosium(3+) trichloride
Molecular Formula
Cl3Dy
InChI Key
BOXVSFHSLKQLNZ-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Dy+3]
Form
Powder
Total Metal Impurities
0.02% max.
Appearance (Color)
White
Residual water
< 100 ppm

Description

It is used as a precursor for the preparation of other dysprosium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text