Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 035692.14
also known as 035692-14
Price (USD)/ Each
1,163.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
1,163.00
Add to cart
Dysprosium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Catalog number035692.14
Price (USD)/ Each
1,163.00
-
Add to cart
Chemical Identifiers
CAS10025-74-8
IUPAC Namedysprosium(3+) trichloride
Molecular FormulaCl3Dy
InChI KeyBOXVSFHSLKQLNZ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Dy+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max.
Appearance (Color)White
Residual water< 100 ppm
FormPowder
It is used as a precursor for the preparation of other dysprosium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only