Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 035693.06
also known as 035693-06
Price (USD)/ Each
343.65
Online exclusive
382.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
343.65
Online exclusive
382.00 
Save 38.35 (10%)
Add to cart
Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number035693.06
Price (USD)/ Each
343.65
Online exclusive
382.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15474-63-2
IUPAC Namedysprosium(3+) triiodide
Molecular FormulaDyI3
InChI KeyRZQFCZYXPRKMTP-UHFFFAOYSA-K
SMILES[I-].[I-].[I-].[Dy+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green
Total Rare Earth Oxide Impurities0.01% max.
FormFlake
Residual water< 100 ppm
It is used in high-intensity metal-halide lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only