Thermo Scientific Chemicals

Erbium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035695.06
5 g, Each
Thermo Scientific Chemicals

Erbium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035695.06
5 g, Each
Quantity
Catalog number: 035695.06
also known as 035695-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13813-42-8
Residual water
< 100 ppm
Appearance (Color)
Purple
Total Rare Earth Oxide Impurities
0.1% max.
Form
Pieces/powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text