Thermo Scientific Chemicals

Gallium(III) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035697.03
1 g, Each
Thermo Scientific Chemicals

Gallium(III) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035697.03
1 g, Each
Quantity
Catalog number: 035697.03
also known as 035697-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-88-9
IUPAC Name
gallium(3+) tribromide
Molecular Formula
Br3Ga
InChI Key
SRVXDMYFQIODQI-UHFFFAOYSA-K
SMILES
[Ga+3].[Br-].[Br-].[Br-]
Appearance (Color)
White
Residual water
< 100 ppm
Form
Powder
Total Metal Impurities
0.002% max.

Description

Gallium bromide is used as a catalyst in the alkylation of aromatic compounds by alkyl bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text