Thermo Scientific Chemicals

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035699.14
25 g, Each
Thermo Scientific Chemicals

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035699.14
25 g, Each
Quantity
Catalog number: 035699.14
also known as 035699-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-91-4
IUPAC Name
gallium(3+) triiodide
Molecular Formula
GaI3
InChI Key
DWRNSCDYNYYYHT-UHFFFAOYSA-K
SMILES
[Ga+3].[I-].[I-].[I-]
Form
Powder
Total Metal Impurities
0.001% max.
Residual water
< 100 ppm
Appearance (Color)
Yellow

Description

Gallium(III) iodide is widely used as lewis acid, a precursor of organogallium reagents. It can also be involved in detection of solar neutrinos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text