Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 035701.03
also known as 035701-03
Price (USD)/ Each
64.65
Online exclusive
71.80 
Save 7.15 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.80 
Save 7.15 (10%)
Add to cart
Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number035701.03
Price (USD)/ Each
64.65
Online exclusive
71.80 
Save 7.15 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-94-3
IUPAC Nameλ²-iron(2+) dichloride
Molecular FormulaCl2Fe
InChI KeyNMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Particle Size-10 Mesh
Total Metal Impurities0.01% max.
FormBeads
Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only