Thermo Scientific Chemicals

Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035701.18
50 g, Each
Thermo Scientific Chemicals

Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035701.18
50 g, Each
Quantity
Catalog number: 035701.18
also known as 035701-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-94-3
IUPAC Name
λ²-iron(2+) dichloride
Molecular Formula
Cl2Fe
InChI Key
NMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Fe++]
Appearance (Color)
White
Particle Size
-10 Mesh
Total Metal Impurities
0.01% max.
Form
Beads

Description

Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text