Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 035702.18
also known as 035702-18
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
Add to cart
Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Catalog number035702.18
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10099-58-8
IUPAC Namelanthanum(3+) trichloride
Molecular FormulaCl3La
InChI KeyICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBeads
Appearance (Color)White
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.1% max.
Particle Size-10 Mesh
Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only