Thermo Scientific Chemicals

Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035702.30
250 g, Each
Thermo Scientific Chemicals

Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035702.30
250 g, Each
Quantity
Catalog number: 035702.30
also known as 035702-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10099-58-8
IUPAC Name
lanthanum(3+) trichloride
Molecular Formula
Cl3La
InChI Key
ICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[La+3]
Form
Beads
Appearance (Color)
White
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.1% max.
Particle Size
-10 Mesh

Description

Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text