Thermo Scientific Chemicals

Lead(II) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035703.03
1 g, Each
Thermo Scientific Chemicals

Lead(II) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035703.03
1 g, Each
Quantity
Catalog number: 035703.03
also known as 035703-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10031-22-8
IUPAC Name
λ²-lead(2+) dibromide
Molecular Formula
Br2Pb
InChI Key
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
SMILES
[Br-].[Br-].[Pb++]
Appearance (Color)
White to pale grey
Form
Beads
Residual water
≤100 ppm
Total Metal Impurities
≤0.001%
Particle Size
-10 Mesh

Description

Lead(II) bromide is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text