Thermo Scientific Chemicals

Magnesium chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 035707.18
50 g, Each
Thermo Scientific Chemicals

Magnesium chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 035707.18
50 g, Each
Quantity
Catalog number: 035707.18
also known as 035707-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7786-30-3
IUPAC Name
magnesium(2+) dichloride
Molecular Formula
Cl2Mg
InChI Key
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES
[Mg++].[Cl-].[Cl-]
Appearance (Color)
White
Form
Bead
Total Metal Impurities
0.1% max.
Particle Size
-10 Mesh

Description

It is widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. It is an important coagulant, an elution buffer for antibody affinity column purifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text