Thermo Scientific Chemicals

Magnesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035709.18
50 g, Each
Thermo Scientific Chemicals

Magnesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035709.18
50 g, Each
Quantity
Catalog number: 035709.18
also known as 035709-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7786-30-3
IUPAC Name
magnesium(2+) dichloride
Molecular Formula
Cl2Mg
InChI Key
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES
[Mg++].[Cl-].[Cl-]
Appearance (Color)
White
Residual water
< 100 ppm
Form
Beads
Particle Size
-10 Mesh
Total Metal Impurities
0.01% max.

Description

Magnesium chloride is generally used as a Lewis acid catalyst. It is used as a flocculating agent, analytical reagent, anesthetic for cephalopods, and also in various magnesium cements, and ceramics. It is used for dust control, and road stabilization. It is used in fire extinguishers, polymerase chain reaction (PCR) and hydrogen storage. It is a precursor for magnesium metal and other magnesium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text