Thermo Scientific Chemicals

Praseodymium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035714.04
2 g, Each
Thermo Scientific Chemicals

Praseodymium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035714.04
2 g, Each
Quantity
Catalog number: 035714.04
also known as 035714-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13536-53-3
IUPAC Name
praseodymium(3+) tribromide
Molecular Formula
Br3Pr
InChI Key
PLKCYEBERAEWDR-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Pr+3]
Appearance (Color)
Green
Form
Powder
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.01% max.

Description

Praseodymium(III) bromide has some of the common uses such as in aircraft engines, motion picture industry, coloring in glasses and enamels, color cubic zirconia to simulate peridot, welder and glass blower goggles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text