Thallium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Thallium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thallium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 035720.18
also known as 035720-18
Price (USD)/ Each
566.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
566.00
Add to cart
Thallium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number035720.18
Price (USD)/ Each
566.00
-
Add to cart
Chemical Identifiers
CAS7789-40-4
IUPAC Nameλ¹-thallanylium bromide
Molecular FormulaBrTl
InChI KeyPGAPATLGJSQQBU-UHFFFAOYSA-M
SMILES[Br-].[Tl+]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
Appearance (Color)Yellow
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Thallium(I) bromide is used in blue sensitive photodetectors. It is also used as a real-time x-ray image sensor. Further, it acts as a standard for elemental thallium. In addition to this, it is a compound semiconductor used in room temperature X- and gamma-ray detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only