Thermo Scientific Chemicals

Thallium(I) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035721.06
5 g, Each
Thermo Scientific Chemicals

Thallium(I) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035721.06
5 g, Each
Quantity
Catalog number: 035721.06
also known as 035721-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-12-0
IUPAC Name
λ¹-thallanylium chloride
Molecular Formula
ClTl
InChI Key
GBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES
[Cl-].[Tl+]
Form
Beads
Total Metal Impurities
0.001% max.
Appearance (Color)
White
Residual water
< 100 ppm
Particle Size
-10 Mesh

Description

It is used as a catalyst in chlorination's and in suntan lamps. The low solubility of TlCl is exploited in chemical synthesis: treatment of metal chloride complexes with TlPF6, gives the corresponding metal hexafluorophosphate derivative. It is used as a tracer in medical imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text