Thermo Scientific Chemicals

Zinc bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035725.22
100 g, Each
Thermo Scientific Chemicals

Zinc bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035725.22
100 g, Each
Quantity
Catalog number: 035725.22
also known as 035725-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7699-45-8
IUPAC Name
zinc(2+) dibromide
Molecular Formula
Br2Zn
InChI Key
VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES
[Zn++].[Br-].[Br-]
Appearance (Color)
White
Particle Size
-10 Mesh
Total Metal Impurities
0.001% max.
Form
Beads

Description

Zinc bromide is an optimal catalyst for stereo specific and regioselective reaction of silacyclopropanes with carbonyl compounds. It is used as a transparent shield against radiation, as a fluid to displace drilling muds in transition phases, as a lewis acid in organic chemistry, colloidon emulsions for photography, a nutritional or functional ingredient in food and oral care products..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text