Thermo Scientific Chemicals

Zinc iodide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035727.14
25 g, Each
Thermo Scientific Chemicals

Zinc iodide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035727.14
25 g, Each
Quantity
Catalog number: 035727.14
also known as 035727-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10139-47-6
IUPAC Name
zinc(2+) diiodide
Molecular Formula
I2Zn
InChI Key
UAYWVJHJZHQCIE-UHFFFAOYSA-L
SMILES
[Zn++].[I-].[I-]
Appearance (Color)
White
Total Metal Impurities
0.005% max.
Residual water
< 100 ppm
Form
Granular

Description

Zinc iodide is used as an x-ray opaque penetrant in industrial radiography to enhance the contrast. It is used as electrode compartments and as a stain in electron microscopy in association with osmium tetroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text