Thermo Scientific Chemicals

Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035730.03
1 g, Each
Thermo Scientific Chemicals

Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035730.03
1 g, Each
Quantity
Catalog number: 035730.03
also known as 035730-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15238-00-3
IUPAC Name
λ²-cobalt(2+) diiodide
Molecular Formula
CoI2
InChI Key
AVWLPUQJODERGA-UHFFFAOYSA-L
SMILES
[Co++].[I-].[I-]
Appearance (Color)
Black
Form
Beads
Residual water
< 100 ppm
Total Metal Impurities
0.001% max.
Particle Size
-10 Mesh

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text